Learn More
Biomatik Corporation Progesterone (PG) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06798
The Progesterone (PG) ELISA Kit allows for quantitative determination of Progesterone in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 1.23-100 ng/mL. Sensitivity: 0.47 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Progesterone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3599