Learn More
Biomatik Corporation Human Programmed Cell Death Protein 1 Ligand 2 (PDL2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06804
The Human Programmed Cell Death Protein 1 Ligand 2 (PDL2) ELISA Kit allows for quantitative determination of Programmed Cell Death Protein 1 Ligand 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BQ51. Cross Activity: No significant cross-reactivity or interference between Programmed Cell Death Protein 1 Ligand 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3604