Learn More
Biomatik Corporation Human Programmed Cell Death Protein 6 Interacting Protein (PDCD6IP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06807
The Human Programmed Cell Death Protein 6 Interacting Protein (PDCD6IP) ELISA Kit allows for quantitative determination of Programmed Cell Death Protein 6 Interacting Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8WUM4. Cross Activity: No significant cross-reactivity or interference between Programmed Cell Death Protein 6 Interacting Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3606