Learn More
Biomatik Corporation Human Prolactin (PRL) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06819
The Human Prolactin (PRL) ELISA Kit allows for quantitative determination of Prolactin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5THQ0. Cross Activity: No significant cross-reactivity or interference between Prolactin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3609