Learn More
Biomatik Corporation Human Prosaposin (PSAP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06850
Encompass_Preferred
The Human Prosaposin (PSAP) ELISA Kit allows for quantitative determination of Prosaposin in Serum, Plasma, Tissue homogenates, Cell lysates, Cerebrospinal fluid, seminal Plasma, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07602. Cross Activity: No significant cross-reactivity or interference between Prosaposin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3620