Learn More
Biomatik Corporation Mouse Protein Disulfide Isomerase A4 (PDIA4) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06904
The Mouse Protein Disulfide Isomerase A4 (PDIA4) ELISA Kit allows for quantitative determination of Protein Disulfide Isomerase A4 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08003. Cross Activity: No significant cross-reactivity or interference between Protein Disulfide Isomerase A4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3641