Learn More
Biomatik Corporation Mouse Receptor Activator of Nuclear Factor Kappa B Ligand (RANkL) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07020
The Mouse Receptor Activator of Nuclear Factor Kappa B Ligand (RANkL) ELISA Kit allows for quantitative determination of Receptor Activator of Nuclear Factor Kappa B Ligand in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O35235. Cross Activity: No significant cross-reactivity or interference between Receptor Activator of Nuclear Factor Kappa B Ligand and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3668