Learn More
Biomatik Corporation Rat Renalase (RNLS) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07072
The Rat Renalase (RNLS) ELISA Kit allows for quantitative determination of Renalase in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.67 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5U2W9. Cross Activity: No significant cross-reactivity or interference between Renalase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3688