Learn More
Biomatik Corporation Rat Renin (REN) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07076
The Rat Renin (REN) ELISA Kit allows for quantitative determination of Renin in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08424. Cross Activity: No significant cross-reactivity or interference between Renin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3691