Learn More
Biomatik Corporation Mouse Resistin Like Beta (RETNLb) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07087
The Mouse Resistin Like Beta (RETNLb) ELISA Kit allows for quantitative determination of Resistin Like Beta in Serum, Plasma and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.63 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q99P86. Cross Activity: No significant cross-reactivity or interference between Resistin Like Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3696