Learn More
Biomatik Corporation Mouse R-Spondin 1 (RSPO1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07146
The Mouse R-Spondin 1 (RSPO1) ELISA Kit allows for quantitative determination of R-Spondin 1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Z132. Cross Activity: No significant cross-reactivity or interference between R-Spondin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3717