Learn More
Biomatik Corporation Human Secreted Phosphoprotein 2 (SPP2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07225
The Human Secreted Phosphoprotein 2 (SPP2) ELISA Kit allows for quantitative determination of Secreted Phosphoprotein 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 28.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13103. Cross Activity: No significant cross-reactivity or interference between Secreted Phosphoprotein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3753