Learn More
Biomatik Corporation Human Selectin, Endothelium (SELE) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07242
The Human Selectin, Endothelium (SELE) ELISA Kit allows for quantitative determination of Selectin, Endothelium in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 39-2,500 pg/mL. Sensitivity: 16 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16581. Cross Activity: No significant cross-reactivity or interference between Selectin, Endothelium and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3762