Learn More
Biomatik Corporation Rat Semaphorin 3A (SEMA3A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07259
Encompass_Preferred
The Rat Semaphorin 3A (SEMA3A) ELISA Kit allows for quantitative determination of Semaphorin 3A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.62-1000 pg/mL. Sensitivity: 6.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q63548. Cross Activity: No significant cross-reactivity or interference between Semaphorin 3A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3775