Learn More
Biomatik Corporation Human Semaphorin 7A (SEMA7A) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07269
The Human Semaphorin 7A (SEMA7A) ELISA Kit allows for quantitative determination of Semaphorin 7A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75326. Cross Activity: No significant cross-reactivity or interference between Semaphorin 7A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3782