Learn More
Biomatik Corporation Mouse Serotonin Transporter (SERT) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07294
The Mouse Serotonin Transporter (SERT) ELISA Kit allows for quantitative determination of Serotonin Transporter in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q60857. Cross Activity: No significant cross-reactivity or interference between Serotonin Transporter and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3791