Learn More
Biomatik Corporation Human Sirtuin 1 (SIRT1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07348
The Human Sirtuin 1 (SIRT1) ELISA Kit allows for quantitative determination of Sirtuin 1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96EB6. Cross Activity: No significant cross-reactivity or interference between Sirtuin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3804