Learn More
Biomatik Corporation Mouse Sirtuin 1 (SIRT1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07349
The Mouse Sirtuin 1 (SIRT1) ELISA Kit allows for quantitative determination of Sirtuin 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q923E4. Cross Activity: No significant cross-reactivity or interference between Sirtuin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3805