Learn More
Biomatik Corporation Human Sirtuin 6 (SIRT6) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07357
The Human Sirtuin 6 (SIRT6) ELISA Kit allows for quantitative determination of Sirtuin 6 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8N6T7. Cross Activity: No significant cross-reactivity or interference between Sirtuin 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3809