Learn More
Biomatik Corporation Human Superoxide Dismutase 1, Soluble (SOD1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07499
The Human Superoxide Dismutase 1, Soluble (SOD1) ELISA Kit allows for quantitative determination of Superoxide Dismutase 1, Soluble in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 26.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00441. Cross Activity: No significant cross-reactivity or interference between Superoxide Dismutase 1, Soluble and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3851