Learn More
Biomatik Corporation Rat Superoxide Dismutase 2, Mitochondrial (SOD2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07503
The Rat Superoxide Dismutase 2, Mitochondrial (SOD2) ELISA Kit allows for quantitative determination of Superoxide Dismutase 2, Mitochondrial in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 31.2-2000 pg/mL. Sensitivity: 14.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07895. Cross Activity: No significant cross-reactivity or interference between Superoxide Dismutase 2, Mitochondrial and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3854