Learn More
Biomatik Corporation Rat Surfactant Associated Protein D (SPD) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07531
The Rat Surfactant Associated Protein D (SPD) ELISA Kit allows for quantitative determination of Surfactant Associated Protein D in Serum, Plasma, Lung lavage fluid and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35248. Cross Activity: No significant cross-reactivity or interference between Surfactant Associated Protein D and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3870