Learn More
Biomatik Corporation Human Synuclein Gamma (SNCg) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07563
The Human Synuclein Gamma (SNCg) ELISA Kit allows for quantitative determination of Synuclein Gamma in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O76070. Cross Activity: No significant cross-reactivity or interference between Synuclein Gamma and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3884