Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Biomatik Corporation Human Thrombin Activatable Fibrinolysis Inhibitor (TAFI) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07622
The Human Thrombin Activatable Fibrinolysis Inhibitor (TAFI) ELISA Kit allows for quantitative determination of Thrombin Activatable Fibrinolysis Inhibitor in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.133 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96IY4. Cross Activity: No significant cross-reactivity or interference between Thrombin Activatable Fibrinolysis Inhibitor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3907