Learn More
Biomatik Corporation Human Thymic Stromal Lymphopoietin (TSLP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07656
The Human Thymic Stromal Lymphopoietin (TSLP) ELISA Kit allows for quantitative determination of Thymic Stromal Lymphopoietin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-1,000 pg/mL. Sensitivity: 26.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q969D9. Cross Activity: No significant cross-reactivity or interference between Thymic Stromal Lymphopoietin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3924