Learn More
Biomatik Corporation Human Thymosin Beta 4 (Tb4) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07666
The Human Thymosin Beta 4 (Tb4) ELISA Kit allows for quantitative determination of Thymosin Beta 4 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.47-200 ng/mL. Sensitivity: 0.91 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P62328. Cross Activity: No significant cross-reactivity or interference between Thymosin Beta 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3928