Learn More
Biomatik Corporation Rat Thyrotropin Releasing Hormone (TRH) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07689
The Rat Thyrotropin Releasing Hormone (TRH) ELISA Kit allows for quantitative determination of Thyrotropin Releasing Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 6.17-500 pg/mL. Sensitivity: 2.43 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01150. Cross Activity: No significant cross-reactivity or interference between Thyrotropin Releasing Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3937