Learn More
Biomatik Corporation Human Tissue Factor (TF) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07696
The Human Tissue Factor (TF) ELISA Kit allows for quantitative determination of Tissue Factor in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13726. Cross Activity: No significant cross-reactivity or interference between Tissue Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3941