Learn More
Biomatik Corporation Rat Tissue Factor Pathway Inhibitor (TFPI) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07703
The Rat Tissue Factor Pathway Inhibitor (TFPI) ELISA Kit allows for quantitative determination of Tissue Factor Pathway Inhibitor in Plasma, Urine, Tissue homogenates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q02445. Cross Activity: No significant cross-reactivity or interference between Tissue Factor Pathway Inhibitor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3946