Learn More
Biomatik Corporation Human Topoisomerase II (TOP2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07768
The Human Topoisomerase II (TOP2) ELISA Kit allows for quantitative determination of Topoisomerase II in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.235 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11388. Cross Activity: No significant cross-reactivity or interference between Topoisomerase II and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3977