Learn More
Biomatik Corporation Triiodothyronine (T3) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07884
The Triiodothyronine (T3) ELISA Kit allows for quantitative determination of Triiodothyronine in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 123.5-10000 pg/mL. Sensitivity: 51.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Triiodothyronine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4025