Learn More
Biomatik Corporation Human Tryptophan-2, 3-dioxygenase (TDO) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07933
The Human Tryptophan-2, 3-dioxygenase (TDO) ELISA Kit allows for quantitative determination of Tryptophan-2, 3-dioxygenase in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P48775. Cross Activity: No significant cross-reactivity or interference between Tryptophan-2, 3-dioxygenase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4042