Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU07947
The Mouse Tumor Necrosis Factor Alpha (TNFa) ELISA Kit allows for quantitative determination of Tumor Necrosis Factor Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P06804. Cross Activity: No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4049