Learn More
Biomatik Corporation Mouse Tyrosine Hydroxylase (TH) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08003
The Mouse Tyrosine Hydroxylase (TH) ELISA Kit allows for quantitative determination of Tyrosine Hydroxylase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24529. Cross Activity: No significant cross-reactivity or interference between Tyrosine Hydroxylase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4073