Learn More
Biomatik Corporation Human Ubiquitin Conjugating Enzyme E2L3 (UBE2L3) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08020
The Human Ubiquitin Conjugating Enzyme E2L3 (UBE2L3) ELISA Kit allows for quantitative determination of Ubiquitin Conjugating Enzyme E2L3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P68036. Cross Activity: No significant cross-reactivity or interference between Ubiquitin Conjugating Enzyme E2L3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4077