Learn More
Biomatik Corporation Human Ubiquitin Cross Reactive Protein (UCRP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08021
The Human Ubiquitin Cross Reactive Protein (UCRP) ELISA Kit allows for quantitative determination of Ubiquitin Cross Reactive Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05161. Cross Activity: No significant cross-reactivity or interference between Ubiquitin Cross Reactive Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4078