Learn More
Biomatik Corporation Human VEGF Co Regulated Chemokine 1 (VCC1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08121
The Human VEGF Co Regulated Chemokine 1 (VCC1) ELISA Kit allows for quantitative determination of VEGF Co Regulated Chemokine 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6UXB2. Cross Activity: No significant cross-reactivity or interference between VEGF Co Regulated Chemokine 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4123