Learn More
Biomatik Corporation Human Visfatin (VF) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08154
The Human Visfatin (VF) ELISA Kit allows for quantitative determination of Visfatin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P43490. Cross Activity: No significant cross-reactivity or interference between Visfatin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4135