Learn More
Biomatik Corporation Human X-Ray Repair Cross Complementing 5 (XRCC5) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08243
The Human X-Ray Repair Cross Complementing 5 (XRCC5) ELISA Kit allows for quantitative determination of X-Ray Repair Cross Complementing 5 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13010. Cross Activity: No significant cross-reactivity or interference between X-Ray Repair Cross Complementing 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4165