Learn More
Biomatik Corporation High Sensitive Human Alpha-2-Macroglobulin (a2M) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08259
The High Sensitive Human Alpha-2-Macroglobulin (a2M) ELISA Kit allows for quantitative determination of Alpha-2-Macroglobulin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 28.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01023. Cross Activity: No significant cross-reactivity or interference between Alpha-2-Macroglobulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4171