Learn More
Biomatik Corporation High Sensitive Mouse Apolipoprotein B (APOB) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08260
The High Sensitive Mouse Apolipoprotein B (APOB) ELISA Kit allows for quantitative determination of Apolipoprotein B in Serum, Plasma and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25-400 ng/mL. Sensitivity: 1.95 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: E9Q414. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4172