Learn More
Biomatik Corporation High Sensitive Human Interleukin 6 (IL6) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08276
The High Sensitive Human Interleukin 6 (IL6) ELISA Kit allows for quantitative determination interleukin 6 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 pg/mL. Sensitivity: 0.35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05231. Cross Activity: No significant cross-reactivity or interference between Interleukin 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4180