Learn More
Biomatik Corporation Sheep Interleukin 1 Beta (IL1b) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08353
The Sheep Interleukin 1 Beta (IL1b) ELISA Kit allows for quantitative determination interleukin 1 Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Ovis aries; Ovine (Sheep). Detection Range: 15.62-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21621. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4197