Learn More
Biomatik Corporation Human Glycated Hemoglobin A1c (HbA1c) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08671
The Human Glycated Hemoglobin A1c (HbA1c) ELISA Kit allows for quantitative determination of Glycated Hemoglobin A1c in Serum, Plasma and Erythrocyte lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 ug/mL. Sensitivity: 4.95 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Glycated Hemoglobin A1c and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4256