Learn More
Biomatik Corporation Rat Procollagen I N-Terminal Propeptide (PINP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08717
The Rat Procollagen I N-Terminal Propeptide (PINP) ELISA Kit allows for quantitative determination of Procollagen I N-Terminal Propeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 246.9-20,000 pg/mL. Sensitivity: 91.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P02454. Cross Activity: No significant cross-reactivity or interference between Procollagen I N-Terminal Propeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4272