Learn More
Biomatik Corporation Human Hemojuvelin (HJV) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08735
The Human Hemojuvelin (HJV) ELISA Kit allows for quantitative determination of Hemojuvelin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1000 ng/mL. Sensitivity: 4.93 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q6ZVN8. Cross Activity: No significant cross-reactivity or interference between Hemojuvelin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4277