Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 15 (FGF15) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08757
The Rat Fibroblast Growth Factor 15 (FGF15) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 15 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 12.5-200 pg/mL. Sensitivity: 4.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q8VI81. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 15 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4287