Learn More
Biomatik Corporation Mouse Erythropoietin (EPO) ELISA Kit (CLIA), 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08774
The Mouse Erythropoietin (EPO) ELISA Kit (CLIA) allows for quantitative determination of Erythropoietin in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.37-1,000 pg/mL. Sensitivity: 0.59 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P07321. Cross Activity: No significant cross-reactivity or interference between Erythropoietin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4290