Learn More
Biomatik Corporation Human Chemokine Like Factor Superfamily 6 (CKLFSF6) ELISA Kit (CLIA), 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08961
The Human Chemokine Like Factor Superfamily 6 (CKLFSF6) ELISA Kit (CLIA) allows for quantitative determination of Chemokine Like Factor Superfamily 6 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.9-5,000 pg/mL. Sensitivity: 2.9 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q9NX76. Cross Activity: No significant cross-reactivity or interference between Chemokine Like Factor Superfamily 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4307