Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 10 (MMP10) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU08979
The Rabbit Matrix Metalloproteinase 10 (MMP10) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 10 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 62.50-4000 pg/mL. Sensitivity: 25.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: G1TEU2. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 10 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4310